Learn More
RAE Systems Inc CALIBRATION MIX
SDP
Catalog No. NC0374078
Encompass_Preferred
Four-Gas Calibration Mix ,58L Aluminum Cylinder (50%LEL / 2.5% VOL CH4,18% O2,10 PPM H2S, 50 PPM CO, BAL N2)
Catalog No. NC0374078 Supplier RAE Systems Inc Supplier No. 6000050070
Four-Gas Calibration Mix ,58L Aluminum Cylinder (50%LEL / 2.5% VOL CH4,18% O2,10 PPM H2S, 50 PPM CO, BAL N2)