Learn More
PCB Congeners Mixture, EPA 8082A, Isooctane, 1 mL
Specifications
Specifications
| Chemical Name or Material | Calibration Mixture |
| CAS | 540-84-1, 12674-11-2, 11096-82-5 |
| CAS Max % | 99.71, 0.14, 0.14 |
| Analyte or Component Names | Isooctane,Aroclor 1016,Aroclor 1260 |
| Element | Isooctane, Aroclor 1016, Aroclor 1260 |
| Solvent or Matrix | Isooctane |
| Quantity | 1 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 1000μg/mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.