Learn More
Cole-Parmer InnoCal NIST-Traceable Calibration; Temperature Baths
Supplier:  Cole-Parmer 1700113
Encompass
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost! NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
Catalog No. NC1907611