Learn More
Heathrow Scientific Calibration Salts,, Potassium Sulfate
Description
- It is recommended to perform water activity calibration if the measurement result of the saturated salt under the same conditions as sample measurement deviates by more than ±0.02* from the reference value
Specifications
Specifications
| Product Type | Calibration Salt |
| Accuracy | 0.973 aw |
| For Use With (Equipment) | Water Activity Meter |