Learn More
LECO Corporation CALIBRATION SAMPLE EDTA 50G
Catalog No. NC9126589
Encompass
Calibration Sample, EDTA, 40.8 to 41.2% Carbon, 5.46 to 5.54% Hydrogen, 9.52 to 9.6% Nitrogen, 50g
Catalog No. NC9126589 Supplier LECO Corporation Supplier No. 502896
Calibration Sample, EDTA, 40.8 to 41.2% Carbon, 5.46 to 5.54% Hydrogen, 9.52 to 9.6% Nitrogen, 50g