Learn More
Randox CALIBRATION SERA LEVEL 2
SDP
Catalog No. NC0270687
Encompass_Preferred
Clinical Chemistry Calibration Serum Level 2. Lyophilised for enhanced stability, Human based serum, Instrument specific target values provided for 42 parameters, Stable to expiry date at +2-8 deg.C, Reconstituted stability of 7 days at +2-8 deg.C or 4 weeks at -20 deg.C.
Catalog No. NC0270687 Supplier Randox Supplier No. CAL2350
Clinical Chemistry Calibration Serum Level 2. Lyophilised for enhanced stability, Human based serum, Instrument specific target values provided for 42 parameters, Stable to expiry date at +2-8 deg.C, Reconstituted stability of 7 days at +2-8 deg.C or 4 weeks at -20 deg.C.