Learn More
Agilent Technologies Semiquantitative Calibration Standard 1, Multi Component Mixture, 100 mL
Catalog No. NC1731111
Encompass
This semi-quantitative calibration standard is designed as a certified reference material for elemental analysis techniques, including ICP-OES, ICP-MS, AAS, MP-AES, and XRF. The multi-component mixture contains 34 elements, each at a concentration of 10 μg/mL, prepared in a 40% aqua regia matrix for stability.
  • Certified reference material manufactured under ISO 17034, ISO/IEC 17025, and ISO 9001 standards.
  • Traceable to NIST SRMs through gravimetric methods and high-performance ICP-OES protocols.
  • Homogeneity confirmed via replicate analysis consistent with ISO Guide 35.
  • Supplied in a 100 mL bottle for precision laboratory use.
  • Requires storage at controlled room temperature, protected from light and moisture.
Catalog No. NC1731111 Supplier Agilent Technologies Supplier No. 51908594
This semi-quantitative calibration standard is designed as a certified reference material for elemental analysis techniques, including ICP-OES, ICP-MS, AAS, MP-AES, and XRF. The multi-component mixture contains 34 elements, each at a concentration of 10 μg/mL, prepared in a 40% aqua regia matrix for stability.
  • Certified reference material manufactured under ISO 17034, ISO/IEC 17025, and ISO 9001 standards.
  • Traceable to NIST SRMs through gravimetric methods and high-performance ICP-OES protocols.
  • Homogeneity confirmed via replicate analysis consistent with ISO Guide 35.
  • Supplied in a 100 mL bottle for precision laboratory use.
  • Requires storage at controlled room temperature, protected from light and moisture.