Learn More
Perkin Elmer US LLC Dual-Detector Calibration Solution for ELAN 9000/6X00, 1,000 mL
Supplier:  Perkin Elmer US LLC N8125010
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 1% HNO3 200 µg/mL: Cd, Cu, Pb, Mg, Rh
Catalog No. 50-904-5604