Learn More
Inorganic Ventures 200.8 CAL SOLUTION 1 125mL
SDP
Catalog No. 50723003
Encompass_Preferred
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Dilution: 1:100, Analyte: Sb and Mo 20 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-003 Supplier Inorganic Ventures Supplier No. 2008CAL1125ML
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Dilution: 1:100, Analyte: Sb and Mo 20 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search