Learn More
Inorganic Ventures 200.8 CAL SOLUTION 2 125mL
SDP
Catalog No. 50723004
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Dilution: 1:100, Analyte: Al, Ba, Cd, Co, Pb, Ni, Ag, Th, V, As, Be, Cr, Cu, Mn, Se, Tl, U, and Zn 20 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-004 Supplier Inorganic Ventures Supplier No. 2008CAL2125ML
Matrix: Nitric Acid (HNO3), Dilution: 1:100, Analyte: Al, Ba, Cd, Co, Pb, Ni, Ag, Th, V, As, Be, Cr, Cu, Mn, Se, Tl, U, and Zn 20 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search