Learn More
Inorganic Ventures 200.7WW CAL SOLUTION 3 125mL
SDP
Catalog No. 50723857
Encompass_Preferred
Inorganic Ventures' Calibration Standard 3 is a 125mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-857 Supplier Inorganic Ventures Supplier No. WWCAL3125ML
Inorganic Ventures' Calibration Standard 3 is a 125mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search