Learn More
Inorganic Ventures CLP CAL SOLUTION 3 125mL
SDP
Catalog No. 50723515
Encompass_Preferred
Inorganic Ventures' Calibration Standard 3 is a 125mLcertified reference materialset in a nitric acid matrix for stability. This arsenic standard isTraceable to NISTand manufactured in accordance with our stringentQuality Assuranceguidelines. One yearExpiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-515 Supplier Inorganic Ventures Supplier No. CLPPCAL3125ML
Inorganic Ventures' Calibration Standard 3 is a 125mLcertified reference materialset in a nitric acid matrix for stability. This arsenic standard isTraceable to NISTand manufactured in accordance with our stringentQuality Assuranceguidelines. One yearExpiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search