Learn More
Inorganic Ventures 200.7WW CAL SOLUTION 4B 125mL
SDP
Catalog No. 50723859
Encompass_Preferred
Inorganic Ventures' Calibration Standard 4B is a 125mL certified reference material set in a nitric acid (tr. hydrofluoric acid) matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-859 Supplier Inorganic Ventures Supplier No. WWCAL4B125ML
Inorganic Ventures' Calibration Standard 4B is a 125mL certified reference material set in a nitric acid (tr. hydrofluoric acid) matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search