Learn More
Electron Microscopy Sciences Calibration Standard Model 70-1D with Cert
SDP
Catalog No. 5034060
Encompass_Preferred
A precision pattern providing accurate calibration in the horizontal plane for very high resolution, nanometer-scale measurements. Period: 70 nm pitch, one-dimensional array. Accurate to +/- 0.25 nm. Refer to calibration certificate for actual pitch. Surface: Silicon Oxide ridges on Silicon, 4x3 mm die. Ridge height (about 35 nm) and width (about 35 nm) are not calibrated.
Catalog No. 50-340-60 Supplier Electron Microscopy Sciences Supplier No. 801271DC
A precision pattern providing accurate calibration in the horizontal plane for very high resolution, nanometer-scale measurements. Period: 70 nm pitch, one-dimensional array. Accurate to +/- 0.25 nm. Refer to calibration certificate for actual pitch. Surface: Silicon Oxide ridges on Silicon, 4x3 mm die. Ridge height (about 35 nm) and width (about 35 nm) are not calibrated.