Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Calibrator 80
SDP

Catalog No. 21377293
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-377-293 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Handle with care. Avoid contact with skin or eyes. Use in a well-ventilated area.
Catalog No. 21-377-293 Supplier Arkray Supplier No. 103400
 Restricted Item
Add to Cart
Add to Cart
Handle with care. Avoid contact with skin or eyes. Use in a well-ventilated area.

Arkray Calibrator 80 provides accurate and reliable calibration for Arkray analyzers, ensuring precise measurement results. Optimized for consistent performance, it supports reproducible laboratory testing and streamlines routine workflows.

  • Reliable Accuracy: Arkray Calibrator 80 ensures precise calibration for Arkray analyzers, delivering dependable results every time.
  • Optimized Performance: Formulated to work seamlessly with Arkray systems, minimizing errors and maximizing efficiency.
  • Time-Saving Convenience: Ready-to-use calibrator reduces preparation time, helping labs operate smoothly and efficiently.
  • Consistent Quality: Each batch meets strict quality standards, ensuring reproducibility and confidence across runs.
  • Enhanced Precision: Supports accurate measurements for multiple assays, improving overall data reliability.
  • Workflow Efficiency: Simplifies lab processes, reduces downtime, and streamlines routine testing operations.
  • Versatile Application: Compatible with a wide range of Arkray assays, making it a flexible choice for diverse laboratory needs.
  • Stable and Dependable: Maintains stability under standard storage conditions, reducing frequent replacement.
  • Minimizes Variability: Supports consistent results across operators and instruments for reliable diagnostics.
  • Trusted Brand: Backed by Arkray's reputation for quality and innovation in clinical diagnostics.

Specifications

Type Solution
Form Liquid

Arkray Calibrator 80 provides accurate and consistent calibration for Arkray analyzers, supporting reliable results and streamlined laboratory workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.