Learn More
Roche Diagnostics Multi-calibrator solution, Calibrator A Bio
Supplier:  Roche Diagnostics 06682189001
Encompass
This product is intended for calibration of quantitative assays of the Cedex Bio and Cedex Bio HT Analyzers for the following tests: Acetate (ACB, ACD) Glucose (GLC2B, GLC2D, GLC3B, GLC3D) Glutamate (GLUB, GLUD) Glycerol (GLYB, GLYD) IgG (IGGLB, IGGHB, IGGHD) Lactate (LAC2B, LAC2D) Phosphate (PHO2B, PHO2D) Total Protein (TP2B, TP2D) Use this calibrator as described in the instructions of the respective assay.
Catalog No. 50-589-435