Learn More
Roche Diagnostics CALIBRATOR C BIO
Supplier:  Roche Diagnostics 07020716001
Encompass
Set C consisting of a multi-calibrator and multi-controls for the Cedex Bio and Cedex Bio HT Systems serve for calibration and QC of the tests for Iron, Calcium, Magnesium
Catalog No. 50-589-451