Learn More
Cole-Parmer Fluke Calibration 9132 Infrared digital calibrator
Supplier:  Cole-Parmer 9091110
Encompass
Check and maintain the accuracy of all your in-line, handheld, or benchtop infrared thermometers with this high-accuracy calibrator. The isothermal target can be controlled in increments of 0.1° from 122 to 932°F (50 to 500°C) by using the front panel controls. Calibration is simple: Set the desired calibration temperature via the front-panel mounted controls, then point your IR thermometer at the target. Compare the readings on the calibrator with the reading on your thermometer and record the difference. For higher precision calibration, use the RTD reference well with an optional RTD thermometer to check the accuracy of the calibrator. For a complete selection of RTD thermometers. Available for sale in the United States and Canada only.
Catalog No. 50-993-446
Add to Cart
Request a Quote