Learn More
Cole-Parmer Fluke 724 Multifunction temperature calibrator
Supplier:  Cole-Parmer 9095000
Encompass
Calibrator takes measurements, simulates, verifies, and calibrates temperature sensors, transmitters, PLCs, and recorders. Ideal for field use—use for maintenance and troubleshooting. Measure and source up to 10 thermocouple types, 7 RTD types, voltage, and resistance. Measure current and power transmitters with 24 VDC loop power supply. Meter performs linearity tests with 25% and 100% stepping; memory stores eight frequently used tests for faster testing. Meter has dual channels and dual LCD—displays measure and source values simultaneously. Backlit display is easy to see even in low-light areas. Calibrator is easy to operate with simple controls and "no menu" operation — no user training required. Available for sale in the United States and Canada only.
Catalog No. 50-993-420