Learn More
Electron Microscopy Sciences pH Buffer Set pH 7 40 ML
SDP
Catalog No. 501929326
Encompass_Preferred
These are pH calibration and buffer solutions to allow you to adjust your testing system to measure your samples' unknown pH more precisely. Always choose a pH 7 buffer plus at least one other pH value close to your expected measurement range. Calibration should be performed before each reading or each set of readings. For best accuracy perform standardizations with fresh buffer solutions. Never pour used buffer back into the bottle. Use buffers directly above and below the expeted value of your sample. Match the temperature of the buffer to the temperature of the samples.
Catalog No. 50-192-9326 Supplier Electron Microscopy Sciences Supplier No. 7208902
These are pH calibration and buffer solutions to allow you to adjust your testing system to measure your samples' unknown pH more precisely. Always choose a pH 7 buffer plus at least one other pH value close to your expected measurement range. Calibration should be performed before each reading or each set of readings. For best accuracy perform standardizations with fresh buffer solutions. Never pour used buffer back into the bottle. Use buffers directly above and below the expeted value of your sample. Match the temperature of the buffer to the temperature of the samples.