Learn More
Cayman Chemical CalIndolhydrochlorIde 10mg
SDP
Catalog No. 502966912
Encompass_Preferred
A phenylalkylamine calcimimetic that facilitates the activation of CaSR, a GPCR that triggers PI-intracellular Ca2+ signaling; stimulates PI hydrolysis and proliferation through human CaSR (pEC50 values are 6.9 and 7.4, respectively) in the presence of calcium
Catalog No. 50-296-6912 Supplier Cayman Chemical Supplier No. 1757510 MG
A phenylalkylamine calcimimetic that facilitates the activation of CaSR, a GPCR that triggers PI-intracellular Ca2+ signaling; stimulates PI hydrolysis and proliferation through human CaSR (pEC50 values are 6.9 and 7.4, respectively) in the presence of calcium