Learn More
MSI Viking Gage MITUTOYO Absolute Digimatic Caliper 0-6
SDP
Catalog No. NC0677350
Encompass_Preferred
The ZERO/ABS key allows the display to be Zero-Set at any slider position along the scale for incremental comparison measurements. This switch will also allow return to the absolute (ABS) coordinate and display of the true position from the origin point (usually jaws-closed point). Data Hold Unit (959143) is optional. Carbide-tipped jaw type calipers are also available. With thumb roller. Supplied in fitted plastic case.
Catalog No. NC0677350 Supplier MSI Viking Gage Supplier No. 50019630
The ZERO/ABS key allows the display to be Zero-Set at any slider position along the scale for incremental comparison measurements. This switch will also allow return to the absolute (ABS) coordinate and display of the true position from the origin point (usually jaws-closed point). Data Hold Unit (959143) is optional. Carbide-tipped jaw type calipers are also available. With thumb roller. Supplied in fitted plastic case.