Learn More
Aviva Systems Biology Corp CALM3 Antibody - N-terminal region (ARP59635_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59635_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CALM3. It was validated on Western Blot. . Alternate names: CALM1, CALM2, PHKD, PHKD3ApplicationWB. Species reactivity: Human, Pig, Rat
Catalog No. 50-286-765