Learn More
Medchemexpress LLC HY-D0801 100mg Medchemexpress, Calmagite CAS:3147-14-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD0801100MG
Encompass_Preferred
Medchemexpress, HY-D0801 100mg Calmagite CAS:3147-14-6 Calmagite is a complexometric indicator which can be used to detect calcium and magnesium in various samples. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3183