Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calmagite
SCI_ED

Catalog No. S25674 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25674 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25674
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 3147-14-6
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) 358.368
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O

Specifications

Linear Formula C17H14N2O5S
Formula Weight 358.38
Packaging Solid
Quantity 5 g
Chemical Name or Material Calmagite
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.