Learn More
Medchemexpress LLC Calmagite | 3147-14-6 | 98.0% | 1 ML
SDP
Catalog No. 5000410275
Encompass_Preferred
Calmagite is a chromomeric reagent utilized for detecting calcium and magnesium in various samples. It is intended for research use only.
  • Chromomeric reagent
  • Used to detect calcium and magnesium in various samples
  • Appearance: solid
  • Color: light brown to black
Catalog No. 50-004-10275 Supplier Medchemexpress LLC Supplier No. HYD080110MM/1ML
Calmagite is a chromomeric reagent utilized for detecting calcium and magnesium in various samples. It is intended for research use only.
  • Chromomeric reagent
  • Used to detect calcium and magnesium in various samples
  • Appearance: solid
  • Color: light brown to black