Learn More
Medchemexpress LLC Calmagite | 3147-14-6 | 98.0% | 500 MG
SDP
Catalog No. 5000410934
Encompass_Preferred
Calmagite is a chromomeric reagent which can be used to detect calcium and magnesium in various samples.
  • Solid appearance
  • Light brown to black color
Catalog No. 50-004-10934 Supplier Medchemexpress LLC Supplier No. HYD0801500MG
Calmagite is a chromomeric reagent which can be used to detect calcium and magnesium in various samples.
  • Solid appearance
  • Light brown to black color