Learn More
Selleck Chemical LLC Calmidazolium chloride
SDP
Catalog No. 503137550
Encompass_Preferred
Calmidazolium chloride (R 24571) is a calmodulin (CaM) antagonist antagonizing CaM-stimulated fraction of rat brain phosphodiesterase and CaM-induced activation of erythrocyte Ca2 -transporting ATPase with IC50s of 0 15 and 0 35 M respectively also shows a CaM-binding Kd value of 3 nM
Catalog No. 50-313-7550 Supplier Selleck Chemical LLC Supplier No. E04835MG
Calmidazolium chloride (R 24571) is a calmodulin (CaM) antagonist antagonizing CaM-stimulated fraction of rat brain phosphodiesterase and CaM-induced activation of erythrocyte Ca2 -transporting ATPase with IC50s of 0 15 and 0 35 M respectively also shows a CaM-binding Kd value of 3 nM