Learn More
Medchemexpress LLC Calmidazolium chloride | 57265-65-3 | MFCD00077679 | 99.9% | 687.70 | C31H23Cl7N2O
SDP
Catalog No. 5000246403
Encompass_Preferred
Calmidazolium chloride is a calmodulin antagonist supplied as a research reagent for in vitro biochemical studies. It is used to inhibit calmodulin-dependent enzymes and calcium-transport processes; reported identifiers include CAS 57265-65-3, molecular formula C31H23Cl7N2O, and molecular weight 687.70.
  • Calmodulin antagonist with high affinity binding to CaM (reported Kd ≈ 3 nM).
  • Inhibits CaM-dependent phosphodiesterase and CaM-induced activation of Ca2+ transport (reported IC50s ≈ 0.15 and 0.35 μM).
  • High reported purity in manufacturer COA (example batch: LCMS 99.94%).
  • Supplied as a solid research chemical with storage recommendations to keep sealed and away from moisture.
Catalog No. 50-002-46403 Supplier Medchemexpress LLC Supplier No. HY10331950MG
Calmidazolium chloride is a calmodulin antagonist supplied as a research reagent for in vitro biochemical studies. It is used to inhibit calmodulin-dependent enzymes and calcium-transport processes; reported identifiers include CAS 57265-65-3, molecular formula C31H23Cl7N2O, and molecular weight 687.70.
  • Calmodulin antagonist with high affinity binding to CaM (reported Kd ≈ 3 nM).
  • Inhibits CaM-dependent phosphodiesterase and CaM-induced activation of Ca2+ transport (reported IC50s ≈ 0.15 and 0.35 μM).
  • High reported purity in manufacturer COA (example batch: LCMS 99.94%).
  • Supplied as a solid research chemical with storage recommendations to keep sealed and away from moisture.