Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calmidazolium chloride | 57265-65-3 | MFCD00077679 | 99.9% | 687.70 g/mol | C31H23Cl7N2O | 1 ML
SDP

Catalog No. 5000244659
Encompass_Preferred

Calmidazolium chloride is a calmodulin antagonist used in biochemical and cellular research to inhibit calmodulin-dependent enzymes and calcium-transport processes. Supplied as a 10 mM solution (1 mL), it is suitable as a research reagent for signaling, enzymology, and cellular assays.

  • High purity (approximately 99.9%).
  • Molecular formula C31H23Cl7N2O.
  • Molecular weight 687.70 g/mol.
  • CAS number 57265-65-3.
  • Calmodulin antagonist with reported Kd of 3 nM.
  • Provided as a small-volume, ready-to-use stock solution for experimental convenience.

Catalog No. 50-002-44659 Supplier Medchemexpress LLC Supplier No. HY10331910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calmidazolium chloride is a calmodulin antagonist used in biochemical and cellular research to inhibit calmodulin-dependent enzymes and calcium-transport processes. Supplied as a 10 mM solution (1 mL), it is suitable as a research reagent for signaling, enzymology, and cellular assays.

  • High purity (approximately 99.9%).
  • Molecular formula C31H23Cl7N2O.
  • Molecular weight 687.70 g/mol.
  • CAS number 57265-65-3.
  • Calmodulin antagonist with reported Kd of 3 nM.
  • Provided as a small-volume, ready-to-use stock solution for experimental convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.