Learn More
Medchemexpress LLC HY-103319 5mg Medchemexpress, Calmidazolium chloride CAS:57265-65-3 Purity:>98%
SDP
Catalog No. 501932241
Encompass_Preferred
Medchemexpress, HY-103319 5mg Calmidazolium chloride CAS:57265-65-3 Calmidazolium chloride (R 24571) is a calmodulin (CaMK) antagonist, antagonizing CaM-dependent phosphodiesterase and calmodulin-induced activation of erythrocyte Ca2+-transporting ATPase with IC 50 s of 0.15 and 0.35 μM, respectively [1] . Also in anti-cancer research [2] . Calmidazolium binds to CaMK with a K d of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2241 Supplier Medchemexpress LLC Supplier No. HY1033195MG
Medchemexpress, HY-103319 5mg Calmidazolium chloride CAS:57265-65-3 Calmidazolium chloride (R 24571) is a calmodulin (CaMK) antagonist, antagonizing CaM-dependent phosphodiesterase and calmodulin-induced activation of erythrocyte Ca2+-transporting ATPase with IC 50 s of 0.15 and 0.35 μM, respectively [1] . Also in anti-cancer research [2] . Calmidazolium binds to CaMK with a K d of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.