Learn More
Echelon Biosciences Research Labs Calmodulin Dependent Protein Kinase II (281-309) - 1 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 893401MG
Encompass_Preferred
Calmodulin Dependent Protein Kinase II (281-309) - 1mg , Met-His-Arg-Gln-Glu-Thr-Val-Asp-Cys-Leu-Lys-Lys-Phe-Asn-Ala-Arg-Arg-Lys-Leu-Lys-Gly-Ala-Ile-Leu-Thr-Thr-Met-Leu-Ala-OH Category/Application: Neuropeptides Hormomes This item is non returnable
Catalog No. 50-190-9127