Learn More
Agilent Technologies CALMODULIN AFFINITY RESIN
Catalog No. 50125262
Encompass
Calmodulin Affinity Resin, pCAL vectors derived from T7 RNA polymerase-based pET vectors, Novel purification tag combines small size, gentle elution and high-affinity binding, Easy tag removal resulting in native protein.
Catalog No. 50-125-262 Supplier Agilent Technologies Supplier No. 214303
Calmodulin Affinity Resin, pCAL vectors derived from T7 RNA polymerase-based pET vectors, Novel purification tag combines small size, gentle elution and high-affinity binding, Easy tag removal resulting in native protein.