Learn More
Medchemexpress LLC Calmodulin binding peptide 1 | 104041-80-7 | 98.68% | 5301.10 | 10 MG
SDP
Catalog No. 5000424158
Encompass_Preferred
Calmodulin Binding Peptide 1 is a high-affinity (pM) CaM-binding peptide derived from smooth muscle myosin light chain kinase (MLCK peptide). The interface of the complex formed by Calmodulin Binding Peptide 1 and Ca2+-CaM can be specifically bound by small-molecule inhibitors, serving as a key target for selective regulation of smooth muscle contraction and development of anti-CaM drugs. In vitro, it binds to the human calmodulin fluorescent biosensor hCaM M124C-mBBr with high affinity and can displace the lower-affinity inhibitor MBC from the biosensor.
  • High-affinity CaM-binding peptide
  • Derived from smooth muscle myosin light chain kinase
  • Target for selective regulation of smooth muscle contraction
  • Useful for development of anti-CaM drugs
  • Binds to human calmodulin fluorescent biosensor with high affinity
Catalog No. 50-004-24158 Supplier Medchemexpress LLC Supplier No. HYP180510MG
Calmodulin Binding Peptide 1 is a high-affinity (pM) CaM-binding peptide derived from smooth muscle myosin light chain kinase (MLCK peptide). The interface of the complex formed by Calmodulin Binding Peptide 1 and Ca2+-CaM can be specifically bound by small-molecule inhibitors, serving as a key target for selective regulation of smooth muscle contraction and development of anti-CaM drugs. In vitro, it binds to the human calmodulin fluorescent biosensor hCaM M124C-mBBr with high affinity and can displace the lower-affinity inhibitor MBC from the biosensor.
  • High-affinity CaM-binding peptide
  • Derived from smooth muscle myosin light chain kinase
  • Target for selective regulation of smooth muscle contraction
  • Useful for development of anti-CaM drugs
  • Binds to human calmodulin fluorescent biosensor with high affinity