Learn More
Bioworld Calmodulin-Coated Microplates, 5 Plate(s)
Supplier:  Bioworld 201400151
Encompass
bioWorld's Calmodulin coated 96-well microplates provide apolystyrene surface with physically adsorbed Calmodulin protein. The Calmodulin Ca ++ binding protein is able to bind proteins mainly with hydrophobic sites in its surface.Applications:• Interactions with proteins involved in glycogen metabolism • Interactions with factors involved in neurotransmission mechanism• Interactions with enzymes involved in the NAD*/NADP* phosphorilation system
Catalog No. 50-197-546