Learn More
Medchemexpress LLC HY-P1479A 1mg Medchemexpress, Calmodulin-Dependent Protein Kinase II(290-309) (acetate) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1479A1MG
Encompass_Preferred
Medchemexpress, HY-P1479A 1mg Calmodulin-Dependent Protein Kinase II(290-309) (acetate) CAS: Calmodulin-Dependent Protein Kinase II (290-309) acetate is a potent CaMK antagonist with an IC50 of 52 nM for inhibition of Ca2+/calmodulin-dependent protein kinase II. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3057