Learn More
CPC Scientific H-Pro-Leu-Arg-Arg-Thr-Leu-Ser-Val-Ala-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP010A
Encompass
ONE-LETTER SEQUENCE: PLRRTLSVAA-NH2MOLECULAR FORMULA: C47H87N17O12MOLECULAR WEIGHT:1082.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: R.B. Pearson et al., JBC, 260, 14471 (1985)
Catalog No. 50-211-0163