Learn More
Bioworld Calmodulin from Bovine Brain, 5 mg
Supplier:  Bioworld 220600223
Encompass_Preferred
Calmodulin from bovine brain Phosphodiesterase 3', 5'-cyclic nucleotide activator from bovine brain Ca2+ binding protein that is required for activation of cyclic nucleotide-dependent phosphodiesterase. It is also a cofactor/activator of nitric oxide synthase, calcineurin, and many kinases including ATPase, myosin light chain kinase, and CAM kinase I, II, and III. It mediates ryanodine receptor activation by cyclic ADP ribose and is involved in intracellular Ca2+ homeostasis. One unit will stimulate 0.008 activated unit of phosphodiesterase, 3\', 5\'-cyclic nucleotide in a 3 ml reaction volume at pH 7.5 and 30 C, to 50% of the maximum activity of the enzyme when saturated with activator, in the presence of 0.01 mM Ca2+. Specifications:40,000 units/mg protein MW: 16.8 kDa Storage: -20 C
Catalog No. 50-253-118
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.