Learn More
Bioworld CALMODULIN SEPAROPOREAGAR
Supplier:  Bioworld 201810012
Encompass
CALMODULIN SEPAROPORE (AGAROSE) 4B: Calmodulin Separopore 4B Affinity matrix with immobilized bovine calmodulin.; For affinity purification of calmodulin-binding proteins including calmodulin-regulated proteins in eukaryotic cells and r...
Catalog No. 50-197-653