Learn More
Aviva Systems Biology Corp CALN1 Antibody - N-terminal region (ARP62436_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62436_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CALN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human CALN1. Alternate names: CABP8ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-287-683