Learn More
Enzo Life Sciences Calnexin polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA860F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of dog calnexin. The sequence is completely conserved in human, mouse and rat. UniProt: P24643. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Quail, Porcine, Monkey, Hamster, Guinea pig, Drosophila, Dog, Chicken, Bovine. Applications: ICC, IF, IHC (PS), IP, WB, Electron microscopy. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-651