Learn More
Enzo Life Sciences Calnexin (lumenal domain) (human), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP865F
Encompass_Preferred
Alternative name: CNX. Purity: ≥85% (SDS-PAGE; Western blot) (full length protein). MW: ~74kDa. Formulation: Liquid. In TRIS containing sodium chloride, 5% glycerol, and 5mM calcium chloride. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7651