Learn More
Aviva Systems Biology Corp NUCB1 Antibody - C-terminal region (P101699_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P101699_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NUCB1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: NucbApplicationIHC. Species reactivity: Dog, Mouse, Rat
Catalog No. 50-014-53811