Learn More
Cayman Chemical CalosobruchusIc AcId 2.5mg
SDP
Catalog No. 502947541
Encompass_Preferred
An insect pheromone; induces copulation release activity in male azuki bean weevils (EC50 = 6.5 ng/insect)
Catalog No. 50-294-7541 Supplier Cayman Chemical Supplier No. 329272.5 MG
Add to Cart
Add to Cart
An insect pheromone; induces copulation release activity in male azuki bean weevils (EC50 = 6.5 ng/insect)