Learn More
Medchemexpress LLC Caloxin 2A1 TFA | 99.9% | 1592.54 | 25 MG
SDP
Catalog No. 5000396706
Encompass_Preferred
Caloxin 2A1 TFA is an extracellular plasma membrane Ca2+-ATPase (PMCA) peptide inhibitor. It does not affect basal Mg2+-ATPase or Na+-K+-ATPase.
  • Extracellular plasma membrane Ca2+-ATPase (PMCA) peptide inhibitor.
  • Does not affect basal Mg2+-ATPase or Na+-K+-ATPase.
  • Inhibits Ca2+-Mg2+-ATPase in human erythrocyte leaky ghosts.
  • Inhibits Ca2+-dependent formation of the 140-kDa acid-stable acylphosphate.
  • Increases airway smooth muscle cells (ASMCs) apoptosis.
Catalog No. 50-003-96706 Supplier Medchemexpress LLC Supplier No. 25MGHYP3278A
Caloxin 2A1 TFA is an extracellular plasma membrane Ca2+-ATPase (PMCA) peptide inhibitor. It does not affect basal Mg2+-ATPase or Na+-K+-ATPase.
  • Extracellular plasma membrane Ca2+-ATPase (PMCA) peptide inhibitor.
  • Does not affect basal Mg2+-ATPase or Na+-K+-ATPase.
  • Inhibits Ca2+-Mg2+-ATPase in human erythrocyte leaky ghosts.
  • Inhibits Ca2+-dependent formation of the 140-kDa acid-stable acylphosphate.
  • Increases airway smooth muscle cells (ASMCs) apoptosis.