Learn More
Cayman Chemical CALP3 (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 503015987
Encompass_Preferred
A calcium-like peptide; truncated version of CALP2; inhibits calcium influx through glutamate receptor channels in cultured rat neocortical neurons (IC50 = 37.25 µM); inhibits excitatory cytotoxicity (IC50 = 50.97 µM) and apoptosis (IC50 = 33.41 µM)
Catalog No. 50-301-5987 Supplier Cayman Chemical Supplier No. 244035 MG
A calcium-like peptide; truncated version of CALP2; inhibits calcium influx through glutamate receptor channels in cultured rat neocortical neurons (IC50 = 37.25 µM); inhibits excitatory cytotoxicity (IC50 = 50.97 µM) and apoptosis (IC50 = 33.41 µM)