Learn More
Enzo Life Sciences Ac-Leu-Leu-Nle-al (5 mg) CAS: 110044-82-1
SDP
Catalog No. NC9756978
Encompass_Preferred
Cysteine protease inhibitor. Alternative Name: ALLN, Calpain inhibitor I, Ac-LL-norleucinal. Formula: C20H37N3O4. MW: 383.5. CAS: 110044-82-1. Purity: ≥95% (HPLC). Sequence N-Acetyl-Leu-Leu-Nle-CHO. Appearance: White powder. Solubility: Soluble in ethanol (5 mg/ml) or DMSO (10 mg/ml). Long Term Storage: -20°C. Handling: Store dry and protect from light.
Catalog No. NC9756978 Supplier Enzo Life Sciences Supplier No. BMLP1200005
Cysteine protease inhibitor. Alternative Name: ALLN, Calpain inhibitor I, Ac-LL-norleucinal. Formula: C20H37N3O4. MW: 383.5. CAS: 110044-82-1. Purity: ≥95% (HPLC). Sequence N-Acetyl-Leu-Leu-Nle-CHO. Appearance: White powder. Solubility: Soluble in ethanol (5 mg/ml) or DMSO (10 mg/ml). Long Term Storage: -20°C. Handling: Store dry and protect from light.