Learn More
Apexbio Technology LLC Calpain Inhibitor II, ALLM 136632-32-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062944
Encompass_Preferred
Calpain inhibitor II (also termed ALLM or CPI-2) is a membrane-permeable inhibitor targeting calpain I calpain II as well as cathepsin L and B Calpain inhibition by this compound can activate apoptosis via caspase-mediated pathways Studies conducted in acute lymphoblastic leukemia (ALL) cell lines (such as ALL-1 RS4 11 JURKAT) and non-Hodgkin s lymphoma (NHL) cells (including RAMOS DAUDI) revealed apoptosis induction at concentrations of 50 100 M Additionally apoptosis triggered by calpain inhibitor II appears independent of tyrosine kinases BTK and LYN Unlike calpain inhibitor I calpain inhibitor II does not influence NF- B signaling nor sensitize tumor cells to TRAIL-induced apoptosis This inhibitor is widely employed in apoptosis-related research and studies exploring calpain-associated cellular pathways
Catalog No. 50-000-62944 Supplier Apexbio Technology LLC Supplier No. A26035.1
Calpain inhibitor II (also termed ALLM or CPI-2) is a membrane-permeable inhibitor targeting calpain I calpain II as well as cathepsin L and B Calpain inhibition by this compound can activate apoptosis via caspase-mediated pathways Studies conducted in acute lymphoblastic leukemia (ALL) cell lines (such as ALL-1 RS4 11 JURKAT) and non-Hodgkin s lymphoma (NHL) cells (including RAMOS DAUDI) revealed apoptosis induction at concentrations of 50 100 M Additionally apoptosis triggered by calpain inhibitor II appears independent of tyrosine kinases BTK and LYN Unlike calpain inhibitor I calpain inhibitor II does not influence NF- B signaling nor sensitize tumor cells to TRAIL-induced apoptosis This inhibitor is widely employed in apoptosis-related research and studies exploring calpain-associated cellular pathways