Learn More
CPC Scientific Z-Val-Phe-CHO 0.5MG
SDP
Catalog No. 502109098
Encompass
Potent and cell permeable calpain I and II inhibitor that reduces capsaicin-mediated cell death in cultured dorsal root ganglionand. This peptide is additionally a gamma-secretase-inhibitor that reduces occurrence of apoptosis in H2O2 and A23187 treated PC12 cells.ONE-LETTER SEQUENCE: Z-VF-CHOMOLECULAR FORMULA: C22H26N2O4MOLECULAR WEIGHT:382.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [88191-84-8], RESEARCH AREA: Apoptosis
Catalog No. 50-210-9098 Supplier CPC Scientific Supplier No. CASP094A
Potent and cell permeable calpain I and II inhibitor that reduces capsaicin-mediated cell death in cultured dorsal root ganglionand. This peptide is additionally a gamma-secretase-inhibitor that reduces occurrence of apoptosis in H2O2 and A23187 treated PC12 cells.ONE-LETTER SEQUENCE: Z-VF-CHOMOLECULAR FORMULA: C22H26N2O4MOLECULAR WEIGHT:382.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [88191-84-8], RESEARCH AREA: Apoptosis