Learn More
Bachem Calpain Inhibitor III
SDP
Supplier:  Bachem 4027129.0100
Encompass_Preferred
Calpain Inhibitor III, Z-Val-Phe-aldehyde, 88191-84-8, C22H26N2O4, Mr 382.46, 100mg. MDL 28170. Application of the calpain inhibitor Z-VF-CHO reduced capsaicin-mediated cell death in cultured dorsal root ganglion neurons. Dong et al. observed that this inhibitor modulated Ab formation by inhibiting the formation of intermediate Ab 1-46 and protecting Ab from degradation.
Catalog No. 50-260-549